Skip to Content
Merck
All Photos(1)

Key Documents

693480

Sigma-Aldrich

Methacrylonitrile-d5

97 atom % D, 97% (CP), contains monomethyl ether hydroquinone (as stabilizer)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D2C=C(CD3)CN
CAS Number:
Molecular Weight:
72.12
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

97 atom % D

Assay

97% (CP)

form

liquid

contains

monomethyl ether hydroquinone (as stabilizer)

refractive index

n20/D 1.399 (lit.)

bp

90-92 °C (lit.)

density

0.858 g/mL at 25 °C

mass shift

M+5

SMILES string

[2H]\C([2H])=C(\C#N)C([2H])([2H])[2H]

InChI

1S/C4H5N/c1-4(2)3-5/h1H2,2H3/i1D2,2D3

InChI key

GYCMBHHDWRMZGG-KPAILUHGSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service