Skip to Content
Merck
All Photos(2)

Key Documents

547352

Sigma-Aldrich

mono-tert-Butyl succinate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3OCO(CH2)2CO2H
CAS Number:
Molecular Weight:
174.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

51-54 °C (lit.)

functional group

carboxylic acid
ester

SMILES string

CC(C)(C)OC(=O)CCC(O)=O

InChI

1S/C8H14O4/c1-8(2,3)12-7(11)5-4-6(9)10/h4-5H2,1-3H3,(H,9,10)

InChI key

PCOCFIOYWNCGBM-UHFFFAOYSA-N

General description

mono-tert-Butyl succinate can be prepared by refluxing a mixture of succinic anhydride and N-hydroxyl succinimide in the presence of dimethylaminopyridine and triethylamine in tert-butanol and toluene.

Application

mono-tert-Butyl succinate may be used in the preparation of:
  • N-succinyl-L,L-diaminopimelic acid (SDAP)
  • H3K9TSu (histone H3 lysine 9 thiosuccinyl) peptide
  • tert-butyl 4-oxo-4-(4-tritylpiperazin-1-yl)butanoate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zinc-selective inhibition of the promiscuous bacterial amide-hydrolase DapE: implications of metal heterogeneity for evolution and antibiotic drug design.
Uda NR, et al.
Metallomics : Integrated Biometal Science, 6(1), 88-95 (2014)
Bin He et al.
Journal of the American Chemical Society, 134(4), 1922-1925 (2012-01-24)
Sirtuins, a class of enzymes known as nicotinamide adenine dinucleotide-dependent deacetylases, have been shown to regulate a variety of biological processes, including aging, transcription, and metabolism. Sirtuins are considered promising targets for treating several human diseases. There are seven sirtuins
Ying Li et al.
American journal of nuclear medicine and molecular imaging, 3(1), 44-56 (2013-01-24)
Arylboronates capture aqueous (18)F-fluoride in one step to afford a highly polar (18)F-labeled aryltrifluoroborate anion ((18)F-ArBF(3) (-)) that clears rapidly in vivo. To date however, there is little data to show that a ligand labeled with a prosthetic (18)F-ArBF(3) (-)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service