535826
2-(4,6-Diphenyl-1,3,5-triazin-2-yl)-5-[(hexyl)oxy]-phenol
Synonym(s):
2,4-Diphenyl-6-[2-hydroxy-4-(hexyloxy)phenyl]-1,3,5-triazine, 2-(2-Hydroxy-4-hexyloxyphenyl)-4,6-diphenyl-1,3,5-triazine, 2-[4-(Hexyloxy)-2-hydroxyphenyl]-4,6-diphenyl-1,3,5-triazine
About This Item
Recommended Products
mp
147-151 °C (lit.)
solubility
methyl methacrylate: 15 mg/mL at 20 °C
methylene chloride: 169 mg/mL at 20 °C
chloroform: 254 mg/mL at 20 °C
toluene: 51 mg/mL at 20 °C
SMILES string
CCCCCCOc1ccc(c(O)c1)-c2nc(nc(n2)-c3ccccc3)-c4ccccc4
InChI
1S/C27H27N3O2/c1-2-3-4-11-18-32-22-16-17-23(24(31)19-22)27-29-25(20-12-7-5-8-13-20)28-26(30-27)21-14-9-6-10-15-21/h5-10,12-17,19,31H,2-4,11,18H2,1H3
InChI key
LEVFXWNQQSSNAC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Allows polycarbonates and polyesters to achieve a higher resistance to weathering than conventional benzotriazole UV absorbers. Low tendency to chelate allows its use in polymer formulations containing catalyst residues.
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service