535788
2-(2H-Benzotriazol-2-yl)-6-dodecyl-4-methylphenol
About This Item
Recommended Products
refractive index
n20/D 1.574 (lit.)
Quality Level
mp
>350 °C (lit.)
solubility
H2O: insoluble <0.01 ppm at 20 °C
acetone, chloroform, ethyl acetate, n-hexane, methylene chloride, and toluene: 50 wt. % at 20 °C
density
0.911 g/mL at 25 °C (lit.)
SMILES string
OC(C(N1N=C2C=CC=CC2=N1)=CC(C)=C3)=C3CCCCCCCCCCCC
InChI
1S/C25H35N3O/c1-3-4-5-6-7-8-9-10-11-12-15-21-18-20(2)19-24(25(21)29)28-26-22-16-13-14-17-23(22)27-28/h13-14,16-19,29H,3-12,15H2,1-2H3
InChI key
VQMHSKWEJGIXGA-UHFFFAOYSA-N
Application
Imparts outstanding light stability and protection efficiency without imparting any yellowness to the protected substrate.
Physical form
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.0 °F - closed cup
Flash Point(C)
112.8 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service