Skip to Content
Merck
All Photos(1)

Key Documents

362913

Sigma-Aldrich

N-Z-L-proline methyl ester

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17NO4
CAS Number:
Molecular Weight:
263.29
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]22/D −36.8°, neat

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.5217 (lit.)

density

1.175 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

COC(=O)[C@@H]1CCCN1C(=O)OCc2ccccc2

InChI

1S/C14H17NO4/c1-18-13(16)12-8-5-9-15(12)14(17)19-10-11-6-3-2-4-7-11/h2-4,6-7,12H,5,8-10H2,1H3/t12-/m0/s1

InChI key

BLQYEDXWEDWCNJ-LBPRGKRZSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service