Skip to Content
Merck
All Photos(1)

Key Documents

337064

Sigma-Aldrich

(Trifluoromethoxy)benzene

99%

Synonym(s):

α,α,α-Trifluoroanisole, Phenyl trifluoromethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OCF3
CAS Number:
Molecular Weight:
162.11
Beilstein:
2043132
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

41.3 mmHg ( 25 °C)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.406 (lit.)

bp

102 °C (lit.)

density

1.226 g/mL at 25 °C (lit.)

functional group

fluoro
phenoxy

SMILES string

FC(F)(F)Oc1ccccc1

InChI

1S/C7H5F3O/c8-7(9,10)11-6-4-2-1-3-5-6/h1-5H

InChI key

GQHWSLKNULCZGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

(Trifluoromethoxy)benzene is an aryl trifluoromethyl ether. (Trifluoromethoxy)benzene can be prepared from 4-chloro-1-(trifluoromethoxy)benzene via hydrogenolysis. Conformation of (trifluoromethoxy)benzene in the gas phase has been studied by electron diffraction and spectroscopy supplemented with ab initio calculations.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemistry in hydrogen fluoride. 7. A novel synthesis of aryl trifluoromethyl ethers.
Feiring AE.
The Journal of Organic Chemistry, 44(16), 2907-2910 (1979)
Trifluoromethoxy benzene in the gas phase studied by electron diffraction and spectroscopy supplemented with ab initio calculations.
Shishkov IF, et al.
Journal of Molecular Structure, 567, 339-360 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service