Skip to Content
Merck
All Photos(2)

Key Documents

309664

Sigma-Aldrich

[1R-(2-endo,3-exo)]-3-Hydroxy-4,7,7-trimethylbicyclo[2.2.1]heptane-2-acetic acid

99%

Synonym(s):

(−)-Isoborneolacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20O3
CAS Number:
Molecular Weight:
212.29
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

optical activity

[α]20/D −18°, c = 1 in chloroform

mp

107-111 °C (lit.)

functional group

carboxylic acid
hydroxyl

SMILES string

CC1(C)[C@@H]2CC[C@@]1(C)[C@H](O)[C@H]2CC(O)=O

InChI

1S/C12H20O3/c1-11(2)8-4-5-12(11,3)10(15)7(8)6-9(13)14/h7-8,10,15H,4-6H2,1-3H3,(H,13,14)/t7-,8+,10+,12-/m0/s1

InChI key

LWVQRKXAZCLJPX-PUJIEXKFSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service