Skip to Content
Merck
All Photos(1)

Key Documents

20026

Sigma-Aldrich

2-tert-Butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine on polystyrene

extent of labeling: ~2.2 mmol/g base loading

Synonym(s):

BEMP on polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: proton reactive

extent of labeling

~2.2 mmol/g base loading

particle size

200-400 mesh

Other Notes

Polymer-bound BEMP

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Chiurchiù et al.
Molecules (Basel, Switzerland), 24(24) (2019-12-19)
Highly functionalized furans are the key scaffolds of many pharmaceuticals and bioactive natural products. Herein, we disclose a new fruitful synthesis of polyfunctionalized furans starting from β-nitroenones and α-functionalized ketones. The protocol involves two steps promoted by solid supported species
R. Schwesinger
Chimia, 39, 269-269 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service