Z763756
BeadBug™ prefilled tubes, 2.0 mL capacity
with 1.0 mm Silica glass beads, acid washed
Synonym(s):
homogenization microtubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106300
NACRES:
NB.22
Recommended Products
material
silica beads
sterility
non-sterile
packaging
pack of 50 ea
manufacturer/tradename
Benchmark Scientific D1031-10
technique(s)
PCR: suitable
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
These prefilled tubes fit the BeadBug™ microtube homogenizer, and come with standard acid-washed glass beads, triple-pure zirconium beads, or stainless steel beads.
The triple-pure beads are molecular biology grade, and are acid washed, heat treated and tested free of nuclease.
The stainless steel beads are acid-washed as well.
All tubes are non-skirted with screw-caps
The triple-pure beads are molecular biology grade, and are acid washed, heat treated and tested free of nuclease.
The stainless steel beads are acid-washed as well.
All tubes are non-skirted with screw-caps
Legal Information
BeadBug is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service