Skip to Content
Merck
All Photos(1)

Documents

Z250473

Sigma-Aldrich

Glass beads

diam. 1.0 mm, for Beadbeating

Synonym(s):

Laboratory Glass Spheres

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.32

packaging

pkg of (net weight 357 g)

diam.

1.0 mm

Looking for similar products? Visit Product Comparison Guide

Application


  • Effects of Temperature, Ionic Strength and Humic Acid on the Transport of Graphene Oxide Nanoparticles in Geosynthetic Clay Liner: This study utilizes glass beads to simulate soil environments in the assessment of nanoparticle mobility, providing essential data for environmental safety evaluations (Liu and Jiang, 2024).

  • Design of closed loop for reproduction of gas-dominated hydrate flow: Glass beads are employed to create a realistic model of gas hydrate flows, aiding in the design of safer and more efficient energy transport systems (Wada et al., 2024).

  • Physicochemical cell disruption of Bacillus sp. for recovery of polyhydroxyalkanoates: Demonstrates the use of glass beads in the mechanical disruption process to optimize the yield of bioplastics, contributing to sustainable materials engineering (Bhat et al., 2024).

Packaging

Glass has a density of 2.5 g/cc and is the most commonly used bead media for ′Beadbeating′. Beads are sold in one pound bottles.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luke T Marr et al.
Methods (San Diego, Calif.), 184, 61-69 (2019-12-13)
The assembly of DNA into nucleosomes and higher order chromatin structures serves not only as a means of compaction but also organizes the genome to facilitate crucial processes such as cell division and regulation of gene expression. Chromatin structure generally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service