Skip to Content
Merck
All Photos(1)

Key Documents

287830

Sigma-Aldrich

Tri(p-tolyl)phosphine

98%

Synonym(s):

TPTP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3C6H4)3P
CAS Number:
Molecular Weight:
304.37
Beilstein:
651045
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings

mp

144-148 °C (lit.)

functional group

phosphine

SMILES string

Cc1ccc(cc1)P(c2ccc(C)cc2)c3ccc(C)cc3

InChI

1S/C21H21P/c1-16-4-10-19(11-5-16)22(20-12-6-17(2)7-13-20)21-14-8-18(3)9-15-21/h4-15H,1-3H3

InChI key

WXAZIUYTQHYBFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Shabbir Hasan et al.
Human genomics, 9, 20-20 (2015-08-20)
Insertion and deletion (indel), a common form of genetic variation, has been shown to cause or contribute to human genetic diseases and cancer. With the advance of next-generation sequencing technology, many indel calling tools have been developed; however, evaluation and
Jingna Si et al.
International journal of molecular sciences, 16(11), 26303-26317 (2015-11-06)
Proteins and RNA interaction have vital roles in many cellular processes such as protein synthesis, sequence encoding, RNA transfer, and gene regulation at the transcriptional and post-transcriptional levels. Approximately 6%-8% of all proteins are RNA-binding proteins (RBPs). Distinguishing these RBPs
Adelaide E Clark et al.
Chemosphere, 137, 33-44 (2015-05-20)
An analytical method has been developed for the pressurized liquid extraction (PLE) of a wide range of semi-volatile organic compounds (SVOCs) from atmospheric particulate matter. Approximately 130 SVOCs from eight compound classes were selected as molecular markers of (1) agricultural
Xiaoqing Wang et al.
Chemosphere, 226, 159-165 (2019-03-31)
Organophosphate flame retardants (OPFRs) have been reported to induce cytotoxicity in a structure-dependent manner. The toxic effects may be due to the damage of biomembrane integrity and/or the interference of membrane signal pathway. In this study, the damages of fifteen
Katherine A Parrish et al.
Physical chemistry chemical physics : PCCP, 21(4), 1689-1699 (2018-09-28)
This study examined the solution-phase exchange reactions of triphenylphosphine (PPh3) ligands on Au8L72+ (L = PPh3) gold clusters with three different tolyl ligands using electrospray ionization mass spectrometry to provide insight into how steric differences in the phosphines influence the

Related Content

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service