Merck
22495-U
Supelco
Férula de columna cargada Supeltex M-4, D.E. columna 1/16 pulgadas
I.D. 1/16 in., 1/16 in. O.D. column, pkg of 10 ea
About This Item
Código UNSPSC:
41121815
NACRES:
SB.54
envase
pkg of 10 ea
temperatura máxima
450 °C
D.I.
1/16 in.
compatibilidad
1/16 in. O.D. column
Descripción general
Ferrules are suitable for the attachment of the column and other glass components to the system. Flexible graphite ferrules withstand high temperatures and are capable of compression.
Supelco has refined the design of these graphite ferrules to provide a clean, sharp profile with minimal flash.
Characteristics:
Seals at one-quarter turn past fingertight. Maximum sealing surface contact, reduced risk of column contamination at installation.
Material Composition: flexible graphite
Maximum Temperature: 450 °C
Información legal
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Walter Jennings
Gas Chromatography with Glass Capillary Columns (2012)