EP5920750003
Adapter for Eppendorf® S-4x Universal Large Rotor
capacity, 1 x 250 mL Corning® bottle, pack of 2
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
Productos recomendados
feature
capacity 1 x 250 mL Corning® bottle
Holds 250 mL tubes
packaging
pack of 2
manufacturer/tradename
Eppendorf® 5920750003 (Global)
technique(s)
centrifugation: suitable
compatibility
for use with Universal-Large bucket
General description
Adapters for various sizes of tubes and bottles for use with Eppendorf S-4x Universal large rotor.
Legal Information
Corning is a registered trademark of Corning, Inc.
Eppendorf is a registered trademark of Eppendorf AG