EP5495511008
Eppendorf® Lid
for use with 5425
About This Item
Productos recomendados
feature
rotor lid
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5495511008
technique(s)
PCR: suitable
centrifugation: suitable
compatibility
for use with 5425
General description
Eppendorf centrifuge 5425/R lid for rotor F-32-0.2-PCR.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG