Documentos clave
EP5495510001
Eppendorf® Rotor F-32x0.2-PCR
for Centrifuge 5425, Eppendorf® 5495510001
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 32 x 0.2 mL PCR tubes
to hold 4 x 8-tube PCR strips
for Centrifuge 5425
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5495510001
parameter
15,060 max. RCF (18,257 xg)
technique(s)
PCR: suitable
centrifugation: suitable
compatibility
for use with centrifuge models 5425
General description
Eppendorf rotor F-32x0.2-PCR for centrifuge 5425/R. Holds up to 4 × PCR strips or 32 × PCR tubes (0.2 mL). Includes rotor lid.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG