Documentos clave
EP022639226
Eppendorf® Adapter
to hold, 1 x 50mL, for 100 mL round bucket in Rotor A-4-38, pack of 2 ea
About This Item
UNSPSC Code:
41103911
material
POM
feature
to hold 1 x 50mL, for 100 mL round bucket in Rotor A-4-38
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 022639226
compatibility
Rotor A-4-38
General description
Adapter, for 1 conical tube 50 mL, 30 mm × 115 mm, for 100 mL round bucket in Rotor A-4-38, 2 pcs.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG