BMSIPD960010TCB
BeadBlaster™ 96 Tungsten Carbide grinding ball
bead diam. 10 mm, Tungsten carbide grinding jar
About This Item
Productos recomendados
material
tungsten carbide
packaging
pack of 2
manufacturer/tradename
Benchmark Scientific IPD9600-10TCB
bead diam.
10 mm
compatibility
Tungsten carbide grinding jar
1 of 4
Este artículo | BMSIPD960015ZOB | BMSIPD960015PTFB | BMSIPD960025TC |
---|---|---|---|
tungsten carbide | (Zirconium Oxide Grinding) | PTFE (with steel core) | tungsten carbide |
pack of 2 | pack of 2 | pack of 2 | pack of 2 |
Tungsten carbide grinding jar | Zirconium Oxide Grinding grinding jar | BeadBlaster™ 96 | BeadBlaster™ 96 |
10 mm | 15 mm | 15 mm | - |
General description
Tungsten carbide grinding media balls owns excellent performance in wear-resistance and resistance to acid and alkali. High hardness can meet the most majority of metal powder crushing and refining.
Application
Features and Benefits
- High density
- Good resistance to acid and alkali
- High hardness for wear resistance