Merck
15-1370
Sigma-Aldrich
Iron(II) oxide
CP
About This Item
Fórmula empírica (notación de Hill):
FeO
Número de CAS:
Peso molecular:
71.84
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade
CP
form
solid
availability
available only in Japan
density
5.7 g/mL at 25 °C (lit.)
storage temp.
15-25°C
SMILES string
O=[Fe]
InChI
1S/Fe.O
InChI key
UQSXHKLRYXJYBZ-UHFFFAOYSA-N
Storage Class
11 - Combustible Solids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Visite la Librería de documentos
Antony George et al.
ACS applied materials & interfaces, 3(9), 3666-3672 (2011-08-16)
A cost-effective and versatile methodology for bottom-up patterned growth of inorganic and metallic materials on the micro- and nanoscale is presented. Pulsed electrodeposition was employed to deposit arbitrary patterns of Ni, ZnO, and FeO(OH) of high quality, with lateral feature
L F Gamarra et al.
Journal of nanoscience and nanotechnology, 10(7), 4145-4153 (2010-12-07)
The magnetic resonance imaging contrast agent, the so-called Endorem colloidal suspension on the basis of superparamagnetic iron oxide nanoparticles (mean diameter of 5.5 nm) coated with dextran, were characterized on the basis of several measurement techniques to determine the parameters
Haiyan Hong et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(31), 8726-8730 (2011-06-17)
Iron oxide coated platinum nanowires (Pt@Fe(2)O(3)NWs) with a diameter of 2.8 nm have been prepared by the oxygen oxidation of FePt NWs in oleylamine. These "cable"-like NWs were characterised by transmission electron microscopy, X-ray diffraction, X-ray photoelectron spectroscopy and X-ray
Hiroaki Kotani et al.
Journal of the American Chemical Society, 133(10), 3249-3251 (2011-02-19)
The photocatalytic formation of a non-heme oxoiron(IV) complex, [(N4Py)Fe(IV)(O)](2+) [N4Py = N,N-bis(2-pyridylmethyl)-N-bis(2-pyridyl)methylamine], efficiently proceeds via electron transfer from the excited state of a ruthenium complex, [Ru(II)(bpy)(3)](2+)* (bpy = 2,2'-bipyridine) to [Co(III)(NH(3))(5)Cl](2+) and stepwise electron-transfer oxidation of [(N4Py)Fe(II)](2+) with 2 equiv
Fe3O4 core/layered double hydroxide shell nanocomposite: versatile magnetic matrix for anionic functional materials.
Liang Li et al.
Angewandte Chemie (International ed. in English), 48(32), 5888-5892 (2009-07-04)