Key Documents
Z740620
Sigma-Aldrich
Aldrich® Essentials petri dish
soda lime glass, autoclavable
About This Item
UNSPSC Code:
41121812
NACRES:
NB.43
material
soda-lime glass
sterility
non-sterile
feature
autoclavable
lid
packaging
pack of 24 ea
dish capacity
~100 mL
binding type
non-treated surface
General description
Clear, soft flint glass Petri dishes.Bottom and top matched. Reusable soda lime glass, can be autoclaved.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)