Documentos clave
Z685119
Ace powder funnel with flat side
top diam. 75 mm, joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
material
borosilicate glass
joint
joint: ST/NS24/40
manufacturer/tradename
Ace Glass
top diam.
75 mm
General description
Ace powder funnel with standard taper joint and one flattened side for easy of use with multi-neck flasks.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)