Merck
Z319600
Büchi® receiving flask
capacity 50 mL, glass
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
glass
round bottom flask
single-neck
joint
joint SJ35/20
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Büchi 00421
capacity
50 mL
General description
These fit all Rotavapor models.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)