Documentos clave
Z219819
Sigma-Aldrich
Single-bulb ball-tube
capacity 500 mL, joint: ST/NS 24/40, w/drip-tip
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
joint
joint: ST/NS24/40
capacity
500 mL
General description
Arranged horizontally outside of Kugelrohr oven, the ball-tubes cool and collect distillate with minimal loss of material.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)