Merck
327026
Sigma-Aldrich
Silver
wire, diam. 0.5 mm, 99.9% trace metals basis
About This Item
Fórmula lineal:
Ag
Número de CAS:
Peso molecular:
107.87
EC Number:
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23
Quality Level
assay
99.9% trace metals basis
form
wire
resistivity
1.59 μΩ-cm, 20°C
diam.
0.5 mm
bp
2212 °C (lit.)
mp
960 °C (lit.)
density
10.49 g/cm3 (lit.)
SMILES string
[Ag]
InChI
1S/Ag
InChI key
BQCADISMDOOEFD-UHFFFAOYSA-N
Application
Ag wire was used to design Ag/AgCl electrodes to measure the diffusion potential of artificial membranes.
Quantity
4 g = 2 m; 20 g = 10 m
pictograms
Environment
signalword
Warning
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Los clientes también vieron
Slide 1 of 9
1 of 9
Silver wire reel, 10m, diameter 0.05mm, annealed, 99.99%
GF95759960
Silver
Silver wire reel, 20m, diameter 0.5mm, annealed, 99.99%
GF65192463
Silver
Silver wire reel, 50m, diameter 0.05mm, annealed, 99.99%
GF69995569
Silver
Silver wire reel, 20m, diameter 0.125mm, annealed, 99.99%
GF02315247
Silver
Silver wire reel, 2m, diameter 2.0mm, annealed, 99.99%
GF28015084
Silver
Silver wire, 0.15 mm diameter, length 5 m, purity 99.99%
GF18621266
Silver
Ion permeability of artificial membranes evaluated by diffusion potential and electrical resistance measurements.
Shlyonsky V.
Advances in Physiology Education, 37.4, 392-400 (2013)
T Prameela Devi et al.
Indian journal of experimental biology, 51(7), 543-547 (2013-08-01)
A total of 75 isolates belonging to five different species of Trichoderma viz., T. asperellum, T. harzianum, T. longibrachiatum, T. pseudokoningii and T. virens were screened for the production of silver nanoparticles. Although all the isolates produced nanoparticles, T. virens
S S Sudha et al.
Indian journal of experimental biology, 51(5), 393-399 (2013-07-05)
Silver nanoparticles is known to have antimicrobial affects. Cyanobacteria isolates from muthupet mangrove includes Aphanothece sp, Oscillatoria sp, Microcoleus sp, Aphanocapsa sp, Phormidium sp, Lyngbya sp, Gleocapsa sp, Synechococcus sp, Spirulina sp with were set in compliance with their cellular
R Nirmala et al.
Journal of nanoscience and nanotechnology, 13(7), 4686-4693 (2013-08-02)
We report on the preparation and characterization of polyurethane (PU) nanofibers containing silver (Ag) nanoparticles were synthesized by using electrospinning. Two different approaches were adopted to incorporate the Ag nanoparticles in to PU nanofibers. In the first approach, a homogeneous
Rui Wang et al.
Journal of nanoscience and nanotechnology, 13(6), 3851-3854 (2013-07-19)
The present studies reveal that silver nanoparticles (AgNPs) can induce apoptosis and enhance radio-sensitivity on cancer cells. In this paper, we mainly investigated the effect of AgNPs on rat glioma C6 cells upon the combination treatment of hyperthermia treatment (HTT).