Skip to Content
Merck
All Photos(1)

Documents

591319

Sigma-Aldrich

Indole-2,4,5,6,7-d5-3-acetic acid

98 atom % D, 98% (CP)

Synonym(s):

2-(2,4,5,6,7-Pentadeuterio-1H-indol-3-yl)acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D5H4NO2
CAS Number:
Molecular Weight:
180.21
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

solid

mp

165-169 °C (lit.)

mass shift

M+5

SMILES string

[2H]c1c([2H])c([2H])c2c(CC(O)=O)c([2H])[nH]c2c1[2H]

InChI

1S/C10H9NO2/c12-10(13)5-7-6-11-9-4-2-1-3-8(7)9/h1-4,6,11H,5H2,(H,12,13)/i1D,2D,3D,4D,6D

InChI key

SEOVTRFCIGRIMH-SNOLXCFTSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingsong Jiao et al.
Journal of experimental botany, 71(19), 5865-5879 (2020-07-11)
Asparagine-linked glycosylation (N-glycosylation) is one of the most important protein modifications in eukaryotes, affecting the folding, transport, and function of a wide range of proteins. However, little is known about the roles of N-glycosylation in the development of stomata in
Luca Narduzzi et al.
Metabolites, 9(12) (2019-12-01)
Hydrophilic Interaction Liquid Chromatography (HILIC) chromatography is widely applied in metabolomics as a complementary strategy to reverse phase chromatography. Nevertheless, it still faces several issues in terms of peak shape and compounds ionization, limiting the automatic de-convolution and data semi-quantification
Chi-Fa Huang et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(33), E6884-E6891 (2017-08-02)
High vein density, a distinctive trait of C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service