Merck
  • N-glycosylation is involved in stomatal development by modulating the release of active abscisic acid and auxin in Arabidopsis.
N-glycosylation is involved in stomatal development by modulating the release of active abscisic acid and auxin in Arabidopsis.
Journal of experimental botany (2020-07-11)
Qingsong Jiao, Tianshu Chen, Guanting Niu, Huchen Zhang, ChangFang Zhou, Zhi Hong
ABSTRACT
Asparagine-linked glycosylation (N-glycosylation) is one of the most important protein modifications in eukaryotes, affecting the folding, transport, and function of a wide range of proteins. However, little is known about the roles of N-glycosylation in the development of stomata in plants. In the present study, we provide evidence that the Arabidopsis stt3a-2 mutant, defective in oligosaccharyltransferase catalytic subunit STT3, has a greater transpirational water loss and weaker drought avoidance, accompanied by aberrant stomatal distribution. Through physiological, biochemical, and genetic analyses, we found that the abnormal stomatal density of stt3a-2 was partially attributed to low endogenous abscisic acid (ABA) and auxin (IAA) content. Exogenous application of ABA or IAA could partially rescue the mutant's salt-sensitive and abnormal stomatal phenotype. Further analyses revealed that the decrease of IAA or ABA in stt3a-2 seedlings was associated with the underglycosylation of β-glucosidase (AtBG1), catalysing the conversion of conjugated ABA/IAA to active hormone. Our results provide strong evidence that N-glycosylation is involved in stomatal development and participates in abiotic stress tolerance by modulating the release of active plant hormones.
Sigma-Aldrich
Indole-2,4,5,6,7-d5-3-acetic acid, 98 atom % D, 98% (CP)
Sigma-Aldrich
Indole-3-acetic acid sodium salt, BioReagent, suitable for plant cell culture, ≥98%