Skip to Content
Merck
All Photos(2)

Key Documents

335657

Sigma-Aldrich

Magnesium methoxide solution

6-10 wt. % in methanol

Synonym(s):

Magnesium dimethanolate, Magnesium dimethoxide, Magnesium methanolate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(OCH3)2
CAS Number:
Molecular Weight:
86.37
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

composition

Mg, 5.5-10.5% EDTA titration

reaction suitability

core: magnesium

concentration

6-10 wt. % in methanol

refractive index

n20/D 1.3381

density

0.816 g/mL at 25 °C

SMILES string

CO[Mg]OC

InChI

1S/2CH3O.Mg/c2*1-2;/h2*1H3;/q2*-1;+2

InChI key

CRGZYKWWYNQGEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in a sol-gel preparation of nanocrystalline MgO with 1.8-35 nm particle sizes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.0 °F - closed cup

Flash Point(C)

7.77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabel Franco Castillo et al.
Journal of materials chemistry. B, 7(41), 6412-6419 (2019-10-24)
Microorganisms such as bacteria, fungi, algae and moulds are highly proficient at colonizing artistic and architectural heritage. The irreparable damage they cause to unique artefacts results in immeasurable cultural and societal losses to our shared cultural heritage, which represent an
Chadwick, A.V. et al.
Chemistry of Materials, 10, 864-864 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service