Merck
Activated N-nitrosocarbamates for regioselective synthesis of N-nitrosoureas.
Journal of medicinal chemistry (1982-02-01)
J Martinez, J Oiry, J L Imbach, F Winternitz
RESUMEN
A practical and convenient method for synthesizing antitumor compounds, N-alkyl-N-nitrosoureas, regioselectively nitrosated on the nitrogen atom bearing the alkyl group is proposed. N-Alkyl-N-nitrosocarbamates are interesting intermediates in these syntheses and yield, by reaction with amino compounds, the regioselectively nitrosated N-alkyl-N-nitrosoureas. As an interesting example, N,N'-bis[(2-chloroethyl)nitrosocarbamoyl]cystamine, a new attractive oncostatic derivative, has been prepared. The cytotoxic activity of these various compounds were tested on L1210 leukemia.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
N-Succinimidyl N-methylcarbamate, ≥97.0% (N)