Merck
Polyol mediated synthesis of ZnO nanoparticles templated by bacterial cellulose.
Carbohydrate polymers (2013-02-13)
Shiyan Chen, Bihui Zhou, Weili Hu, Wen Zhang, Na Yin, Huaping Wang
RESUMEN
Zinc oxide nanoparticles have been successfully synthesized through a facile polyol method using bacterial cellulose (BC) as a template. BC membrane was used as a host matrix to introduce quantitatively Zn(2+) ions and then as nanoreactors to fabricate ZnO nanoparticles by hydrolysis of zinc acetate in a polyol medium. The influence of the concentration of zinc acetate and hydrolytic time on the morphologies and size of ZnO nanoparticles were investigated. The results indicated that the uniform spherical ZnO nanoparticles were incorporated into BC fibers. The resulting nanocomposites show good mechanical properties and high photocatalytic activity in the degradation of methyl orange.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Zinc acetate dihydrate, ACS reagent, ≥98%
Sigma-Aldrich
Zinc acetate dihydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc acetate dihydrate, reagent grade
Sigma-Aldrich
Zinc acetate dihydrate, 99.999% trace metals basis
Sigma-Aldrich
Zinc acetate dihydrate, puriss., E 650, 99-102%
Sigma-Aldrich
Zinc acetate dihydrate, JIS special grade, ≥99.0%