Merck
Helical polymer as mimetic enzyme catalyzing asymmetric aldol reaction.
Macromolecular rapid communications (2012-02-10)
Dongyue Zhang, Chonglei Ren, Wantai Yang, Jianping Deng
RESUMEN
This Communication reports optically active helical substituted polyacetylenes which solely catalyzed asymmetric Aldol reaction between cyclohexanone and p-nitrobenzaldehyde; more importantly the helical structures are found to play crucial roles in the asymmetric catalysis, with a remarkable yield and ee (both up to 80%). A synergic effect is observed between the helical structures in the polymer main chains and the pendent prolinamide moieties for successfully catalyzing the asymmetric reaction. The role of the helical polymer backbones is further verified by tuning the relative helical structure content.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
4-Nitrobenzaldehyde, 98% (GC)
Sigma-Aldrich
Cyclohexanone, ACS reagent, ≥99.0%
Sigma-Aldrich
Cyclohexanone, 99.8%
Sigma-Aldrich
Cyclohexanone, ReagentPlus®, 99.8%
Sigma-Aldrich
Cyclohexanone, puriss. p.a., ≥99.5% (GC)
Supelco
Cyclohexanone, analytical standard
Sigma-Aldrich
Cyclohexanone, JIS special grade, ≥99.0%
Supelco
Cyclohexanone, Selectophore, ≥99.5%
Sigma-Aldrich
Cyclohexanone, SAJ first grade, ≥98.0%