Merck
Chronic Activation of Liver X Receptor Sensitizes Mice to High Cholesterol Diet-Induced Gut Toxicity.
Molecular pharmacology (2018-07-27)
Wojciech G Garbacz, Hirdesh Uppal, Jiong Yan, Meishu Xu, Songrong Ren, Donna B Stolz, Min Huang, Wen Xie
RESUMEN
Cholesterol is essential for numerous biologic functions and processes, but an excess of intracellular cholesterol can be toxic. Intestinal cholesterol absorption is a major determinant of plasma cholesterol level. The liver X receptor (LXR) is a nuclear receptor known for its activity in cholesterol efflux and reverse cholesterol transport. In this study, we uncovered a surprising function of LXR in intestinal cholesterol absorption and toxicity. Genetic or pharmacologic activation of LXRα-sensitized mice to a high-cholesterol diet (HCD) induced intestinal toxicity and tissue damage, including the disruption of enterocyte tight junctions, whereas the same HCD caused little toxicity in the absence of LXR activation. The gut toxicity in HCD-fed LXR-KI mice may have been accounted for by the increased intestinal cholesterol absorption and elevation of enterocyte and systemic levels of free cholesterol. The increased intestinal cholesterol absorption preceded the gut toxicity, suggesting that the increased absorption was not secondary to tissue damage. The heightened sensitivity to HCD in the HCD-fed LXRα-activated mice appeared to be intestine-specific because the liver was not affected despite activation of the same receptor in this tissue. Moreover, heightened sensitivity to HCD cannot be reversed by ezetimibe, a Niemann-Pick C1-like 1 inhibitor that inhibits intestinal cholesterol absorption, suggesting that the increased cholesterol absorption in LXR-activated intestine is mediated by a mechanism that has yet to be defined.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Isotiocianato de fluoresceína-dextrano, average mol wt 4,000, (FITC:Glucose = 1:250)
Sigma-Aldrich
Anti-β-actina, anticuerpo monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Ezetimibe, ≥98% (HPLC)