Merck
  • Improved method for the determination of 12 non-nutritive sweeteners and monitoring in various foods using liquid chromatography-tandem mass spectrometry.
Improved method for the determination of 12 non-nutritive sweeteners and monitoring in various foods using liquid chromatography-tandem mass spectrometry.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2018-06-15)
Ho Soo Lim, EunA Choi, Ju Young Hwang, Gunyoung Lee, Sang Soon Yun, MeeKyung Kim
RESUMEN
An improved and highly sensitive method was developed and validated for the determination of 12 (7 permitted and 5 non-permitted in Korea) non-nutritive sweeteners in various foods using liquid chromatography-electrospray ionisation-tandem mass spectrometry. The chromatographic separation was performed on an Xbridge BEH C18 column (3 mm × 100 mm, 2.5 μm) with gradient elution using 10 mM ammonium acetate in water and 10 mM ammonium acetate in methanol. Sample preparation consisted of simple dilution, homogenisation, centrifugation and purification with a C18 cartridge prior to analysis. The relative matrix effect (%ME) was within ±20% for all sweeteners. The method also showed good linearity (R2 > 0.99). The limit of detection and limit of quantification values in sample were in the range of 0.02-2.66 and 0.06-8.05 mg kg-1, respectively. The recoveries at three concentration levels ranged between 80% and 119%, with relative standard deviation values below 10%. In addition, the expanded uncertainties determined for 12 sweeteners in 5 different food matrices were confirmed to be <14%. Finally, the method was successfully applied to the analysis of sweeteners in 681 food samples purchased in Korea, Australia and Turkey. These results demonstrate that the method is suitable for the simultaneous determination of multiple-sweeteners in a variety of foods.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium N-cyclohexylsulfamate, ≥99.0% (T)