Key Documents
RAD190
radiello Barcode Labels
pk of 198
About This Item
UNSPSC Code:
41104008
NACRES:
NB.24
packaging
pk of 198
manufacturer/tradename
Radiello Code 190
technique(s)
passive air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
General description
Each self-adhesive label allows users to record sampling date/time and displays a unique barcode for the unmistakable identification of the cartridge adsorbent during sampling, desorption, and analysis.
Legal Information
radiello is a trademark of Institi Clinici Scientifici Maugeri
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Lot/Batch Number
Lo sentimos, en este momento no disponemos de COAs para este producto en línea.
Visite la Librería de documentos
Benoît Oury et al.
Journal of occupational and environmental hygiene, 3(10), 547-557 (2006-08-16)
Diffusive sampling is particularly suited to determine time-weighted average volatile organic compound (VOC) concentration in occupational hygiene and environmental air monitoring. The purpose of this study was to measure the sampling rate variation of four different samplers in a special
P Bruno et al.
Waste management (New York, N.Y.), 27(4), 539-544 (2006-05-23)
Nuisance caused by odors is one of the most important problems for waste management plants. To control an odor nuisance, it must first be quantified. The analytical difficulties in odor measurements are related to the high number of volatile components
Francesc A Esteve-Turrillas et al.
Journal of chromatography. A, 1216(48), 8549-8556 (2009-10-27)
A new procedure for the passive sampling in air of benzene, toluene, ethylbenzene and xylene isomers (BTEX) is proposed. A low-density polyethylene layflat tube filled with a mixture of solid phases provided a high versatility tool for the sampling of
P Lovreglio et al.
Giornale italiano di medicina del lavoro ed ergonomia, 33(2), 117-124 (2011-07-30)
To study the validity of urinary benzene as a biomarker of low and very low exposure to this toxicant, as compared with t,t-muconic acid (t,t-MA) and S-phenylmercapturic acid (SPMA), also taking into account the influence of cigarette smoking and co-exposure
Alberto Sturaro et al.
Environmental monitoring and assessment, 164(1-4), 93-100 (2009-04-04)
This paper reports the results of a long-term monitoring of benzene, toluene, ethylbenzene, xylenes (BTEX), phenol and cresols in the air of Padua during a wide period of the year 2007 using two radial passive samplers (Radiello system) equipped with
Artículos
Learn the how the diffusive air sampler works to see if it’s the right fit for you.
Learn the how the diffusive air sampler works to see if it’s the right fit for you.
Protocolos
Learn how to assemble the supporting plate of your radiello air sampler in the lab before the sampling campaign to save time in the field.
Contenido relacionado
Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.
Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.