Documentos clave
58062-U
Aparato de filtración de fase móvil 2 Supelco®
Sinónimos:
Supelco Apparatus for Mobile Phase Filtration
About This Item
Código UNSPSC:
41115700
NACRES:
SB.52
En este momento no podemos mostrarle ni los precios ni la disponibilidad
descripción
connects to aspiration line yes
Nivel de calidad
envase
pkg of 1
Categorías relacionadas
Descripción general
Connects to aspiration line
Includes:
  • 250 mL glass reservoir
  • S/T 34/45 funnel base
  • S/T 34/45 1000 mL flask and glass cap
  • Clamp
  • Stainless steel holder and screen
  • 10 PTFE gaskets
  • 50 Nylon 66 filters (47 mm, 0.45 μm pores).
Productos recomendados
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Información legal
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Artículos
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Questions
  1. Buenas tardes, ¿vendéis este equipo por piezas? Necesito reponer la pieza en forma de vaso del equipo de vacío, pero el resto del equipo está intacto.
    1 answer
    1. Yes, the spare parts for this product are sold separately. The cup-shaped part is referred to as the glass reservoir, located on the top of the apparatus. The product number for this item is 58063. Please see the link below to review this product:
      https://www.sigmaaldrich.com/product/supelco/58063