Documentos clave
56142
Conector de pequeño calibre Omnifit®
Three-way valve connector
About This Item
UNSPSC Code:
12000000
En este momento no podemos mostrarle ni los precios ni la disponibilidad
description
PTFE cones included
technique(s)
LPLC: suitable
bore
0.8 mm
General description
Omnifit® Small Bore Connectors offer the unique advantage of producing a very low dead volume connection with tubing of various diameters, through an O-ring compression system, a cone over tubing system or a void-filling cone with a standard gripper.
Features and Benefits
Cono de PTFE incluido.
Legal Information
Omnifit is a registered trademark of Diba Industries
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)