Key Documents
27109-U
Supelco
Crimp Seal with PTFE/Silicone/PTFE Septum
seal diam. 11 mm, red PTFE/silicone/red PTFE septum, pkg of 1000 ea
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
red PTFE/silicone/red PTFE septum
silver aluminum seal
feature
closure type crimp seal
packaging
pkg of 1000 ea
seal diam.
11 mm
fitting
thread for 11 mm
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)