Merck
2631705
Supelco
Inlet Liner, for Packed GC (wool packed)
wool packed, pkg of 5 ea
About This Item
UNSPSC Code:
12000000
packaging
pkg of 5 ea
L × O.D. × I.D.
112 mm × 6.0 mm × 3.0 mm
General description
Deactivated (silanized) glass inlet liner for packed injection ports.
  • Fits PerkinElmer (AutoSystem)
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)