Z381861
Ultramicro pipette tips
Microcapillary, flat tip, maximum volume 10 μL, non-sterile
About This Item
Productos recomendados
material
colorless
polypropylene
polypropylene
sterility
non-sterile
packaging
pack of 200 ea (Bulk packed)
manufacturer/tradename
Sorenson 1735
maximum volume
10 μL
Categorías relacionadas
General description
Low capacity improves accuracy with small sample volumes by reducing dead space.
Application
For loading sequencing gels as thin as 0.2 mm.
Other Notes
Browse all pipette tips