Documentos clave
Z367737
Low-profile roller
extra roller
About This Item
Código UNSPSC:
41103811
NACRES:
NB.73
fabricante / nombre comercial
IBI ACCRLSR10
L × An × Al
362 mm × 381 mm × 60 mm
Descripción general
Weather-proof roller operates on benchtop, in dry or humid incubators, in cold rooms to 0°C, and even in ovens to 65°C. Use for cell culture with standard roller bottles and in filter blot applications with hybridization bottles (below). Unit is supplied with six gear-driven rollers, to rotate three large bottles at variable speed, 0.25−8 rpm. Rollers can be moved closer together to accommodate small bottles and tubes, and up to five more rollers can be added; as many as thirty 10 × 75 mm tubes can be turned at 2−67 rpm. Unit is powered by a cool-running 24 VDC motor. Thin, flat AC power cord (US only) reaches external outlet without damaging incubator or refrigerator gasket.
Características y beneficios
Contains One replacement roller for the IBI Low Profile Roller.
The replacement roller fits directly into the Low Profile Roller unit without any adjustments or assembly. Snaps right into place.
  • Comes complete and ready for installation
  • Black rubber roll with hex head ends for installation
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)