Documentos clave
TT38440SB-EP
Eppendorf twin.tec® PCR Plates 384
skirted, PCR clean, volume 40 μL, colorless wells, blue plate, pack of 25
About This Item
UNSPSC Code:
41106300
material:
blue plate
colorless wells
polypropylene wells
volume:
40 μL
working volume:
40 μL
feature:
skirt
PCR clean
skirted
material
blue plate
colorless wells
polypropylene wells
well size
384
feature
skirt
PCR clean
skirted
packaging
pack of 25
manufacturer/tradename
Eppendorf® 951020737 (NA)
technique(s)
PCR: suitable
volume
40 μL
working volume
40 μL
General description
The Eppendorf twin.tec PCR plate combines extremely thin-walled polypropylene wells for optimal heat transfer to the sample, with robust polycarbonate frames for high rigidity. With raised well rims, twin.tec plates enable highly effective sealing to reduce the risk of cross-contamination.
Features and Benefits
  • Optimized rim for barcodes and automated work processes
  • StackableWithin SBS footprint recommendationsI
  • deal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler X50 and Mastercycler nexus
  • 40 µL max. well volume
  • Ideal for quantitative real-time PCR as well – OptiTrack matrix for faster sample identification and fewer pipetting errors
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)