Merck
SML3497
Sigma-Aldrich
LY345899
≥95% (HPLC)
Sinónimos:
5,6,7,8-Tetrahydro-N5,N10-carbonylfolic acid, LY 345899, LY-345899, LY354899, N-[4-(3-Amino-1,2,5,6,6a,7-hexahydro-1,9-dioxoimidazo[1,5-f]pteridin-8(9H)-yl)benzoyl]-L-glutamic acid
About This Item
Fórmula empírica (notación de Hill):
C20H21N7O7
Número de CAS:
Peso molecular:
471.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Quality Level
assay
≥95% (HPLC)
form
(Powder or Lyophilized powder or film)
storage condition
desiccated
color
white to beige
storage temp.
-10 to -25°C
SMILES string
O=C1N2C3=C(NC(N)=NC3=O)NCC2CN1C4=CC=C(C=C4)C(N[C@H](C(O)=O)CCC(O)=O)=O
Biochem/physiol Actions
LY345899 (LY354899) is a methylenetetrahydrofolate dehydrogenase inhibitor (MTHFD1 IC50 = 96 nM with 30 μM folitixorin and 400 μM NADP+; MTHFD2 IC50 = 663 nM with 5 μM folitixorin and 250 μM NAD+) that exerts antiproliferation activity against cancer cells by targeting the folate cycle. Co-administer LY345899 with the dihydrofolate reductase (DHFR) inhibitor methotrexate (MTX) overcomes AML MTX resistance (50 mg/kg) in mice in vivo (10 mg/kg q.o.d. i.p.).
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Visite la Librería de documentos
Xinxin Ren et al.
Cancer research, 82(1), 60-74 (2021-11-13)
Metabolic reprogramming by oncogenic signaling is a hallmark of cancer. Hyperactivation of Wnt/β-catenin signaling has been reported in hepatocellular carcinoma (HCC). However, the mechanisms inducing hyperactivation of Wnt/β-catenin signaling and strategies for targeting this pathway are incompletely understood. In this
J L Tonkinson et al.
The Journal of pharmacology and experimental therapeutics, 287(1), 315-321 (1998-10-09)
5,6,7,8-Tetrahydro-N5,N10-carbonylfolic acid (LY354899) has been demonstrated to inhibit the dehydrogenase activity of C1-tetrahydrofolate synthase. This compound was only moderately antiproliferative toward CCRF-CEM lymphocytic leukemia cells in culture, but induced apoptosis after long incubation times. Slightly greater potency was observed in
Crystal Structure of the Emerging Cancer Target MTHFD2 in Complex with a Substrate-Based Inhibitor.
Cancer Research, 77, 937-948 (2017)
Nadilly Bonagas et al.
Nature cancer, 3(2), 156-172 (2022-03-02)
The folate metabolism enzyme MTHFD2 (methylenetetrahydrofolate dehydrogenase/cyclohydrolase) is consistently overexpressed in cancer but its roles are not fully characterized, and current candidate inhibitors have limited potency for clinical development. In the present study, we demonstrate a role for MTHFD2 in
Josep Tarragó-Celada et al.
Cancers, 13(3) (2021-01-28)
With most cancer-related deaths resulting from metastasis, the development of new therapeutic approaches against metastatic colorectal cancer (mCRC) is essential to increasing patient survival. The metabolic adaptations that support mCRC remain undefined and their elucidation is crucial to identify potential