Key Documents
OH83033964
Ohaus® Conductivity Meter
Model ST300C
About This Item
UNSPSC Code:
41115609
material
epoxy shaft
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 83033964
conductivity measuring range
0.00-199.9 mS/cm
temperature measuring range
0-100 °C
battery power
4 AAA, 250 hr life
General description
Easy-to-use and accurate with a simple calibration process, automatic temperature compensation and fast results.
Application
Conductivity, total dissolved solids (TDS) measurement
Features and Benefits
Automatic temperature compensation, adjustable temperature coefficient
Legal Information
Ohaus is a registered trademark of Ohaus Corp.