Key Documents
LIDFA451817-EP
Lid for Eppendorf(R) FA-45-18-17 Rotor
aerosol-tight, for 5418/ 5418 R, aluminum
About This Item
UNSPSC Code:
41103913
material
aluminum
feature
aerosol-tight
manufacturer/tradename
Eppendorf® 022652087 (NA)
Eppendorf® 5418708001 (EU)
technique(s)
centrifugation: suitable
compatibility
for use with 5418 R
for use with 5418
Categorías relacionadas
Legal Information
Eppendorf is a registered trademark of Eppendorf AG