HS14522
Aluminium weighing dish
D × H 101 mm × 12 mm , large
About This Item
UNSPSC Code:
41121812
NACRES:
NB.35
Productos recomendados
manufacturer/tradename
Heathrow Scientific HS14522
D × H
101 mm × 12 mm , large
General description
Designed for moisture determination balances.
- Weigh dishes are FDA/food-grade compliant
- Disposable
- Slightly tapered walls for stacking
- Pack of 50