Key Documents
GE80-1106-89
Glass Plates
U-frame, H × W × D 125 mm × 260 mm × 0.5 mm, pack of 2 ea
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
packaging
pack of 2 ea
manufacturer/tradename
Cytiva 80110689
H × W × D
125 mm × 260 mm × 0.5 mm
Storage Class