Documentos clave
EPM1289-0010
Test Tube Rack, for Eppendorf® New Brunswick Innova® and Eppendorf® Excella® Shakers
size medium, holds 60 tubes
About This Item
Código UNSPSC:
41122804
NACRES:
NB.73
tamaño
medium
360x80x129 mm
Características
holds 60 tubes
fabricante / nombre comercial
Eppendorf® M1289-0010 (Global)
Categorías relacionadas
Descripción general
Test Tube Racks have an adjustable angle and tube height ­support. Made from stainless-steel with inserts to help retain tubes and ­prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Información legal
Eppendorf is a registered trademark of Eppendorf AG
Excella is a registered trademark of Eppendorf, Inc. 
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)