Documentos clave
EP5825718003
SBS adapter, for 1 plate with frame in SBS format
for 1 plate with frame in SBS format, pkg of 2 ea
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
feature
for 1 plate with frame in SBS format
packaging
pkg of 2 ea
manufacturer/tradename
Eppendorf® 5825718003
General description
For Use With (Equipment): Rotors A-2-MTP and A-2-DWP
Legal Information
Eppendorf is a registered trademark of Eppendorf AG