Documentos clave
EP5195000079
Eppendorf® TransferTip (ES)
sterile, set of 25 ea
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
sterility:
sterile
packaging:
set of 25 ea
manufacturer/tradename:
5195000079
Eppendorf®
sterility
sterile
packaging
set of 25 ea
manufacturer/tradename
5195000079
Eppendorf®
General description
Eppendorf® is a top company in the life science field. They manufacture and sell tools, materials, and services for handling liquids, samples, and cells in laboratories around the world. Their wide range of products is used in research labs, at universities, and companies.
Application
Eppendorf®TransferTip® (ES) has been used for the microinjection of embryonic stem cells(ESCs) into the blastocysts of rats.
Features and Benefits
  • 15 μm inner diameter.
  • 20° tip angle.
  • Rigid flange, length 1 mm.
  • Heat-formed spike eases penetration.
  • Sterilized via validated gamma irradiation.
  • Capillary safe for optimum protection.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
TransferTip is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Lot/Batch Number
Lo sentimos, en este momento no disponemos de COAs para este producto en línea.
Visite la Librería de documentos
Elizabeth C Bryda et al.
Methods in molecular biology (Clifton, N.J.), 2631, 355-370 (2023-03-31)
The availability of reliable germline competent rat embryonic stem cell (ESC) lines that can be genetically manipulated provides an important tool for generating new rat models. Here we describe the process for culturing rat ESCs, microinjecting the ESCs into rat