Documentos clave
EP3122612006
Eppendorf® Locking Clip
for 8-channel pipette
About This Item
UNSPSC Code:
41121517
NACRES:
NB.01
feature
for 8-channel pipette
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® 3122612006
General description
Accessory for Eppendorf Xplorer/Xplorer plus
Locking clip
Available for 8- and 12-channel pipettes
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)