Key Documents
EP022638657
Wide Neck Bottle for Eppendorf® A-4-81 Rotor
square, capacity 500 mL, pack of 2
Sinónimos:
Lid for Eppendorf® FA-45-48-11 Rotor
About This Item
UNSPSC Code:
41121703
material
polypropylene bottle
square
feature
with cap
packaging
pack of 2
manufacturer/tradename
Eppendorf® 022638657 (US)
parameter
3250 × g max. RCF
capacity
500 mL
Categorías relacionadas
Features and Benefits
  • Made of polypropylene
  • Offers resistance to centrifugation forces, withstanding speeds up to 3,250 x g
  • The bottle is autoclavable, ensuring convenient sterilization for repeated use
Legal Information
Eppendorf is a registered trademark of Eppendorf AG