Documentos clave
CLS4218
Corning® Step-R Repeating Pipettor Adapter for 25 mL Syringe Tips
non-sterile
Sinónimos:
repeater adapter
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
material
red polypropylene
sterility
non-sterile
feature
Autoclavable at 121°C, 25 minutes
packaging
pack of 1 ea
manufacturer/tradename
Corning 4218
General description
Suitable for use with 25 mL syringe tips.
Legal Information
Corning is a registered trademark of Corning, Inc.